missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CRYAB Protein

A cDNA sequence encoding the CRYAB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11518-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937508

  • £2975.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CRYAB Protein
Research Use Only
Recombinant Protein
E. coli
MDIAIHHPWI RRPFFPFHSP SRLFDQFFGE HLLESDLFST ATSLSPFYLR PPSFLRAPSW IDTGLSEMRL EKDRFSVNLD VKHFSPEELK VKVLGDVIEV HGKHEERQDE HGFISREFHR KYRIPADVDP LTITSSLSSD GVLTVNGPRK QVSGPERTIP ITREEKPAVA AAPKK
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
Untagged
The Mouse CRYAB protein solution contains 20mM Tris-HCl buffer pH 8 and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers