missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Complement Component C5a Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 2150-C5-025/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18166570

  • £340.16 / 25µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse Complement Component C5a Protein is derived from E. coli. The Recombinant Mouse Complement Component C5a Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
9.0 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
15139
Complement Component C5a
E. coli-derived mouse Complement Component C5a protein Asn679-Arg755,with an N-terminal Met
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers