missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Complement C1q subcomponent subunit A Protein
A DNA sequence encoding the Mus musculus (Mouse) Complement C1q subcomponent subunit A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5735-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031598.2 | |
Complement C1q subcomponent subunit A Protein | |
Research Use Only | |
C1QA | |
Recombinant Protein | |
E. coli | |
EDVCRAPNGK DGAPGNPGRP GRPGLKGERG EPGAAGIRTG IRGFKGDPGE SGPPGKPGNV GLPGPSGPLG DSGPQGLKGV KGNPGNIRDQ PRPAFSAIRQ NPMTLGNVVI FDKVLTNQES PYQNHTGRFI CAVPGFYYFN FQVISKWDLC LFIKSSSGGQ PRDSLSFSNT NNKGLFQVLA GGTVLQLRRG DEVWIEKDPA KGRIYQGTEA DSIFSGFLIF PSA | |
Greater than 90% as determined by SDS-PAGE. |
12259 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |