missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Accession Number | Q62507 |
| Concentration | LYOPH |
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized from PBS |
| Gene ID (Entrez) | 1690 |
| Quantity | 10 μg |
| Storage Requirements | Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.01 EU/μg purified protein (LAL test; Lonza). |
| Gene Alias | coagulation factor C (Limulus polyphemus homolog); cochlin, coagulation factor C homolog, cochlin (Limulus polyphemus), COCH-5B2COCH5B2, cochlin, DFNA9 |
| Gene Symbol | COCH |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?