missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CMA1 / Chymase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CMA1 / Chymase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5845-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034910.1 | |
CMA1 / Chymase 1 Protein | |
Research Use Only | |
CMA1 | |
Recombinant Protein | |
E. coli | |
IIGGTECIPH SRPYMAYLEI VTSENYLSAC SGFLIRRNFV LTAAHCAGRS ITVLLGAHNK TSKEDTWQKL EVEKQFLHPK YDENLVVHDI MLLKLKEKAK LTLGVGTLPL SANFNFIPPG RMCRA | |
Greater than 90% as determined by SDS-PAGE. |
17228 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |