missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CLF-1/CLC Complex Protein

Product Code. 18709216
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18709216

Brand: R&D Systems™ 10088CL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

His-tag

The Recombinant Mouse CLF-1/CLC Complex Protein is derived from CHO. The Recombinant Mouse CLF-1/CLC Complex Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9JM58 (CLF-1), Q9QZM3 (CLC)
For Use With (Application) Bioactivity
Molecular Weight (g/mol) 67 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse CLF-1/CLC Complex protein, (N-terminus) Mouse CLF-1 (Gly40-Gly425) Accession &Num; Q9JM58 SGGGGSGGGGSGGGGS Mouse CLC (Leu28-Phe225) Accession &Num; Q9QZM3 HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity CLF-1/CLC Complex
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.