missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CKAP4/p63 Protein

Product Code. 18737418
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18737418

Brand: R&D Systems™ 9734CK050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse CKAP4/p63 Protein is derived from NS0. The Recombinant Mouse CKAP4/p63 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAH25522
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 10970
Molecular Weight (g/mol) 53 kDa
Name 63 kDa membrane protein, CLIMP-63, cytoskeleton-associated protein 4, ERGIC-63, MGC99554, P63, transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediatecompartment, type-II transmembrane protein p63
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse CKAP4/p63 protein Val109-Ile575,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.