missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Chymotrypsin-like elastase family member 2A Protein
A DNA sequence encoding the Mus musculus (Mouse) Chymotrypsin-like elastase family member 2A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5817-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031945.1 | |
Chymotrypsin-like elastase family member 2A Protein | |
Research Use Only | |
Cela2a | |
Recombinant Protein | |
E. coli | |
VVGGQEATPN TWPWQVSLQV LSSGRWRHNC GGSLVANNWV LTAAHCLSNY QTYRVLLGAH SLSNPGAGSA AVQVSKLVVH QRWNSQNVGN GYDIALIKLA SPVTLSKNIQ TACLPPAGTI LPRNYVCYVT GWGLLQTNGN SPDTLRQGRL LVVDYATCSS ASWWGSSVKS SMVCAGGDGV TSSCNGDSGG PLNCRASNGQ WQVHGIVSFG SSLGCNYPRK PSVFTRVSNY IDWINSVMAR N | |
Greater than 90% as determined by SDS-PAGE. |
13706 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |