missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CHST7 Protein

Product Code. 18331338
Change view
Click to view available options
:
20ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18331338 20ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331338 Supplier R&D Systems™ Supplier No. 5108ST020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer sulfate from PAPS to N-acetyl-D-glucosamine. The specific activity is >25 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9EP78
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 56548
Molecular Weight (g/mol) M.W. (Observed): 55-60 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Name Carbohydrate Sulfotransferase 7/CHST7
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse Carbohydrate Sulfotransferase 7/CHST7 protein Ser34-Val484, with an N-terminal 6-His tag Accession # Q9EP78
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Chst7
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Carbohydrate Sulfotransferase 7/CHST7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.