missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Chitinase 3-like 1/YKL-40 Protein

Product Code. 18364118
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364118 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364118 Supplier R&D Systems™ Supplier No. 2649CH050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of FaDu human squamous cell carcinoma cells. When 5 x 104 cells per well are added to rmCHI3-L1 coated plate, cell adhesion is enhanced in a dose dependent manner after 30 minutes at 37 °C. The ED50 for this effect is 0.8-3.2 μg/mL.

Optimal concentrations should be determined by each laboratory for each application.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q61362
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1116
Molecular Weight (g/mol) M.W. (Observed): 41 kDa, reducing conditions; M.W. (theoretical): 41.6 kDa
Name Chitinase 3-like 1
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse Chitinase 3-like 1 protein Tyr22-Ala381, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Chil1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Chitinase 3-like 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.