missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDNF Protein is fully biologically active when compared to standard. It is able to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons when immobilized at 5 - 30 μg/mL on a nitrocellulose-coated microplate.
Specifications
Specifications
| Accession Number | Q8CC36 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 441549 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 18.5 kDa |
| Quantity | 1mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of CDNF as determined by LAL method. |
| Gene Alias | arginine-rich, mutated in early stage tumors-like 1, ARMETL1, ARMET-like protein 1, cerebral dopamine neurotrophic factor, Conserved dopamine neurotrophic factor |
| Gene Symbol | CDNF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?