missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CD8 alpha/beta Heterodimer Protein, CF
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its ability of the immobilized protein to support the adhesion of Jurkat human acute T cell leukemia cells. The ED50 for this effect is 0.750-6.00 μg/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 925 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 33-37 kDa & 45-51 kDa, under reducing conditions., MolecularWeight-theroretical: 28 kDa (CD8 alpha) & 27 kDa (CD8 beta) |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived mouse CD8 protein Mouse CD8 alpha (Lys28-Tyr196) (N-terminus) Accession # NP_033987.1 Acidic Tail 6-His tag Mouse CD8 beta (Leu22-Thr175) (N-terminus) Accession # P10300.1 Basic Tail FLAG (DYKDDDDK) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction