missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD74 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD74, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5795-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001036070.1 | |
CD74 Protein | |
Research Use Only | |
Cd74 | |
Recombinant Protein | |
E. coli | |
QQQGRLDKLT ITSQNLQLES LRMKLPKSAK PVSQMRMATP LLMRPMSMDN MLLGPVKNVT KYGNMTQDHV MHLLTRSGPL EYPQLKGTFP ENLKHLKNSM DGVNWKIFES WMKQWLLFEM SKNSLEEKKP TEAPPKVLTK CQEEVSHIPA VYPGAFRPKC DENGNYLPLQ CHGSTGYCWC VFPNGTEVPH TKSRGRHNCS EPLDMEDLSS GLGVTRQELG QVTL | |
Greater than 90% as determined by SDS-PAGE. |
16149 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |