missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD45 Protein

Product Code. 18379427
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379427 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379427 Supplier R&D Systems™ Supplier No. 114CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When 0.5 μg/mL of rmCD45 is captured onto a Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ) coated plate, the concentration of rmGalectin-1 that produces 50% of the optimal binding is found to be approximately 0.45-1.8 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001104786
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5788
Molecular Weight (g/mol) M.W. (Observed): 140-160 kDa, reducing conditions; M.W. (theoretical): 59.5 kDa
Name CD45
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD45 protein Gln24-Lys564 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Ptprc
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein CD45
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.