missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD300d Fc Chimera Protein

Product Code. 18756316
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18756316

Brand: R&D Systems™ 10310LM050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 or IFN-gamma secretion by human T cells. The ED50 for this effect is 1 to 10 μg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number BAF46249.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 100131439
Molecular Weight (g/mol) 44 kDa
Name CD300 molecule-like family member d, CD300d, CD300LD, CLM-5, CMRF35-A4, immune receptor CD300d, MAIR-IV
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse CD300d protein

Mouse CD300d (Asn20-Leu172) Accession # BAF46249.1IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <1.0 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.