missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD27/TNFRSF7 Fc Chimera Protein

Product Code. 18375607
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375607 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375607 Supplier R&D Systems™ Supplier No. 574CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit the proliferation of mouse T cells induced by CD27 Ligand and anti-CD3. The ED50 for this effect is 0.75-3 μg/mL in the presence of 10 μg/mL (100 μL/well) Recombinant Mouse CD27 Ligand/TNFSF7 (Catalog # 783-CL ) and anti-CD3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P41272.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 939
Molecular Weight (g/mol) M.W. (Observed): 60-66 kDa, reducing conditions; M.W. (theoretical): 44.7 kDa (monomer)
Name CD27/TNFRSF7 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD27/TNFRSF7 protein Mouse CD27 (Thr21-Arg182) (N-terminus) Accession # P41272.1 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd27
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein CD27/TNFRSF7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.