missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD27 Ligand/TNFSF7 Protein

Product Code. 18359647
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18359647 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 50µg
18177590 Lyophilized from a 0.2 μm filtered solution in PBS. 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18359647 Supplier R&D Systems™ Supplier No. 783CL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to stimulate mouse T cell proliferation in the presence of anti-CD3. The ED50 for this effect is 1.25-5.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q05A52
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 970
Molecular Weight (g/mol) M.W. (Observed): 30 kDa, reducing conditions; M.W. (theoretical): 19.2 kDa
Name CD27 Ligand/TNFSF7
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD27 Ligand/TNFSF7 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse CD27 Ligand (Gln47 - Pro195) (N-terminus) Accession # Q05A52 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd70
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein CD27 Ligand/TNFSF7/CD70
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.