missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD207 / Langerin Protein

Product Code. 16021411
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16021411 200 μg 200µg
16001411 10 μg 10µg
16041411 50 μg 50µg
16091401 100 μg 100µg
16031411 500 μg 500µg
16011411 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16021411 Supplier enQuireBio™ Supplier No. QP7887ec200ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) CD207 / Langerin, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_659192.2
Gene ID (Entrez) 246278
Name CD207 / Langerin Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Cd207
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His
Sequence QAVFYPRLMG KILDVKSDAQ MLKGRVDNIS TLGSDLKTER GRVDDAEVQM QIVNTTLKRV RSQILSLETS MKIANDQLQI LTMSWGEVDS LSAKIPELKR DLDKASALNT KVQGLQNSLE NVNKLLKQQS DILEMVARGW KYFSGNFYYF SRTPKTWYSA EQFCISRKAH LTSVSSESEQ KFLYKAADGI PHWIGLTKAG SEGDWYWVDQ TSFNKEQSRR FWIPGEPNNA GNNEHCANIR VSALKCWNDG PCDNTFLFIC KRPYVQTTE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.