missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD200R1 Protein
A cDNA sequence encoding the CD200R1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11313-10ug
Additional Details : Weight : 0.01000kg
Specifications
57781 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPTDKNQTT QNNSSSPLTQ VNTTVSVQIG TKALLCCFSI PLTKAVLITW IIKLRGLPSC TIAYKVDTKT NETSCLGRNI TWASTPDHSP ELQISAVTLQ HEGTYTCETV TPEGNFEKNY DLQVLVPPEV TYFPEKNRSA VCEAMAGKPA AQISWSPDGD CVTTSESHSN GTVTVRSTCH WEQNNVSDVS CIVSHLTGNQ SLSIELSRGG NQSLRPLEPK SCDKTHTCPP CPAPELLGGP SVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQ QGNVFSCSVM HEALHNHYTQ KSLSLSPGKH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
CD200R1 Protein | |
Research Use Only | |
Cd200r1 | |
Mouse | |
His | |
CD200R1 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |