missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD20 / MS4A1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD20 / MS4A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6387-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031667.1 | |
CD20 / MS4A1 Protein | |
Research Use Only | |
MS4A1 | |
Recombinant Protein | |
E. coli | |
ILNMTLSHFL KMRRLELIQT SKPYVDIYDC EPSNSSEKNS PSTQYCNSIQ SVFLGILSAM LISAFFQKLV TAGIVENEWK RMCTRSKSNV VLLSAGEKNE QTIKMKEEII ELSGVSSQPK NEEEIEIIPV QEEEEEEAEI NFPAPPQEQE SLPVENEIAP | |
Greater than 90% as determined by SDS-PAGE. |
12482 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |