missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD155/PVR Fc Chimera Protein

Product Code. 18746618
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18746618

Brand: R&D Systems™ 9670CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Mouse CD96 (Catalog # 5690-CD) is coated at 1 μg/mL (100 μL/well), the concentration of Recombinant Mouse CD155/PVR Fc Chimera that produces 50% optimal binding response is 6-36 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_081790
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5817
Molecular Weight (g/mol) M.W. (Observed): 92-108 kDa, reducing conditions; M.W. (theoretical): 62 kDa
Name CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD155/PVR protein Mouse CD155/PVR (Asp29-Leu348) (N-terminus) Accession # NP_081790 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD155/PVR
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.