missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse CCL9/10/MIP-1 gamma Protein

Product Code. 18407861 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407861 5 μg 5µg
18409001 1 mg 1mg
18442672 100 μg 100µg
18410252 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18407861 Supplier Novus Biologicals™ Supplier No. NBP2350865ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Macrophage Marker Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human neutrophils is in a concentration range of 0.1-10 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51670, P51670, P51670
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Molecular Weight (g/mol) 11.6 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of CCL9/10/MIP-1 gamma as determined by LAL method.
Gene Symbol CCL9/10
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Chemokines and Cytokines
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein CCL9/10/MIP-1 gamma
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.