missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL6/C10 protein is fully biologically active when compared to standard. The biologically active determined by a chemotaxis bioassay using human CCR1 transfected murine BaF3 cells is in a concentration range of 10-100 ng/mL
Specifications
Specifications
| Accession Number | P27784, P27784, P27784 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, pH 8.0, 500 mM NaCl. |
| Gene ID (Entrez) | 20305 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 10.7 kDa |
| Quantity | 1mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of CCL6/C10 as determined by LAL method. |
| Gene Alias | c10, C-C motif chemokine 6, MRP, MRP-1, Scy, Scya6 |
| Gene Symbol | Ccl6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?