missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL6 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human T lymphocytes is in a concentration range of 1.0-10 ng/mL.
Specifications
Specifications
| Accession Number | P30882, P30882, P30882 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 30 % Acetonitrile and 0.1 % TFA. |
| Gene ID (Entrez) | 6352 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 7.9 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of CCL5/RANTES as determined by LAL method. |
| Gene Alias | CCL5, chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES/CCL5, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES |
| Gene Symbol | CCL5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?