missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL27 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human peripheral blood lymphocytes is in a concentration range of 10-100 ng/mL.
Specifications
Specifications
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, pH 7.4, with 5% trehalose. |
| Gene ID (Entrez) | 10850 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 10.9 kDa |
| Quantity | 1mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of CCL27/CTACK as determined by LAL method. |
| Gene Alias | ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK/CCL27, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 |
| Gene Symbol | CCL27 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?