missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) Protein

Product Code. 18307427
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18307427 Lyophilized from a 0.2μm filtered solution in PBS. 25µg
18127010 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18307427 Supplier R&D Systems™ Supplier No. 1017MP025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) is derived from E. coli. The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 56221
Molecular Weight (g/mol) 10.3 kDa
Name CCL24/Eotaxin-2/MPIF-2 aa 27-119
Quantity 25 μg
Source E. coli-derived mouse CCL24/Eotaxin-2/MPIF-2 protein Val27-Val119
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.