missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1017-MP-025

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18127010

  • £197.62 / 25µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) Protein is derived from E. coli. The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
10.3 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
56221
CCL24/Eotaxin-2/MPIF-2 aa 27-119
E. coli-derived mouse CCL24/Eotaxin-2/MPIF-2 protein Val27-Val119
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions