missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Accession Number | O88430, O88430, O88430 |
| For Use With (Application) | PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 2x PBS, 0.02% Tween-20, pH 7.0. |
| Gene ID (Entrez) | 6367 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 7.8 kDa |
| Quantity | 1mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of CCL22/MDC as determined by LAL method. |
| Gene Alias | A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, CCL22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC/CCL22, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 |
| Gene Symbol | CCL22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?