missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CCL22/MDC Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca: R&D Systems™ 439-MD-025
Detalles adicionales : Weight : 0.00970kg
Descripción
The Recombinant Mouse CCL22/MDC Protein is derived from E. coli.Especificaciones
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
7.8 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
20299 | |
CCL22/MDC | |
E. coli-derived mouse CCL22/MDC protein Gly25-Ser92 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |