missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CCL20/MIP-3 alpha Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£291.57
Specifications
Gene ID (Entrez) | 20297 |
---|---|
Molecular Weight (g/mol) | 7.9 kDa |
Name | CCL20/MIP-3 alpha |
Quantity | 25μg |
Source | E. coli-derived mouse CCL20/MIP-3 alpha protein Ala27-Met96 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18300398
|
R&D Systems™
760-M3-025 |
£291.57
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18117750
|
R&D Systems™
760-M3-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£291.57
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse CCL20/MIP-3 alpha Protein is derived from E. coli. The Recombinant Mouse CCL20/MIP-3 alpha Protein has been validated for the following applications: Bioactivity.Specifications
20297 | |
CCL20/MIP-3 alpha | |
E. coli-derived mouse CCL20/MIP-3 alpha protein Ala27-Met96 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |