missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL2 / MCP-1 / MCP1 Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_035463.1 |
---|---|
Gene ID (Entrez) | 20296 |
Name | CCL2 / MCP-1 / MCP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966149
|
enQuireBio™
QP5449-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946149
|
enQuireBio™
QP5449-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936149
|
enQuireBio™
QP5449-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976149
|
enQuireBio™
QP5449-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956149
|
enQuireBio™
QP5449-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_035463.1 | |
CCL2 / MCP-1 / MCP1 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
THP-1 chemotaxis significantly induced by less than 100 ng/ml of Mouse CCL2 / MCP-1 / MCP1. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
20296 | |
Research Use Only | |
CCL2 | |
Recombinant Protein | |
E. coli | |
QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN | |
Greater than 95% as determined by SDS-PAGE. |