missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cathepsin G Protein
A DNA sequence encoding the Mus musculus (Mouse) Cathepsin G, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5887-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031826.1 | |
Cathepsin G Protein | |
Research Use Only | |
Ctsg | |
Recombinant Protein | |
E. coli | |
IIGGREARPH SYPYMAFLLI QSPEGLSACG GFLVREDFVL TAAHCLGSSI NVTLGAHNIQ MRERTQQLIT VLRAIRHPDY NPQNIRNDIM LLQLRRRARR SGSVKPVALP QASKKLQPGD LCTVAGWGRV SQSRGTNVLQ EVQLRVQMDQ MCANRFQFYN SQTQICVGNP RERKSAFRGD SGGPLVCSNV AQGIVSYGSN NGNPPAVFTK IQSFMPWIKR TMRRFAPRYQ RPANSLSQAQ T | |
Greater than 90% as determined by SDS-PAGE. |
13035 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |