missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse CAMK2N1 Protein

Product Code. 18258361 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18258361

Brand: Novus Biologicals™ NBP25957750UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-78 of Mouse CAMK2N1 The Recombinant Mouse CAMK2N1 Protein is derived from E. coli. The Recombinant Mouse CAMK2N1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl (pH8.5) containing 1mM DTT, 0.1M NaCl, 10% glycerol
Gene ID (Entrez) 55450
Molecular Weight (g/mol) 10.9 kDa
Name Recombinant Mouse CAMK2N1 Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias Calcium/Calmodulin Dependent Protein Kinase II Inhibitor 1, Calcium/Calmodulin-Dependent Protein Kinase II Inhibitor 1, CaMKII Inhibitory Protein Alpha, CaMKIINalpha, CaMKIIN-Alpha, PRO1489
Gene Symbol VAMK2N1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.