missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse c-Myc OncoProtein
A DNA sequence encoding the Mus musculus (Mouse) c-Myc Oncoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6398-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001170823.1 | |
c-Myc OncoProtein | |
Research Use Only | |
MYC | |
Recombinant Protein | |
E. coli | |
MPLNVNFTNR NYDLDYDSVQ PYFICDEEEN FYHQQQQSEL QPPAPSEDIW KKFELLPTPP LSPSRRSGLC SPSYVAVATS FSPREDDDGG GGNFSTADQL EMMTELLGGD MVNQSFICDP DDETFIKNII IQDCMWSGFS AAAKLVSEKL ASYQAARKDS TSLSPARGHS VCSTSSLYLQ DLTAAASECI DPSVVFPYPL NDSSSPKSCT SSDSTAFSPS SDSLLSSESS PRASPEPLVL HEETPPTTSS DSEEEQEDEE EIDVVSVEKR QTPAKRSESG SSPSRGHSKP PHSPLVLKRC HVSTHQHNYA APPSTRKDYP AAKRAKLDSG RVLKQISNNR KCSSPRSSDT EENDKRRTHN VLERQRRNEL KRSFFALRDQ IPELENNEKA PKVVILKKAT AYILSIQADE HKLTSEKDLL RKRREQLKHK LEQLRNSGA | |
Greater than 90% as determined by SDS-PAGE. |
17869 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |