missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C-C motif chemokine 25 Protein
A DNA sequence encoding the Mus musculus (Mouse) C-C motif chemokine 25, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8649-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033164.1 | |
C-C motif chemokine 25 Protein | |
Research Use Only | |
Ccl25 | |
Recombinant Protein | |
E. coli | |
GAFEDCCLGY QHRIKWNVLR HARNYHQQEV SGSCNLRAVR FYFRQKVVCG NPEDMNVKRA MRILTARKRL VHWKSASDSQ TERKKSNHMK SKVENPNSTS VRSATLGHPR MVMMPRKTNN | |
Greater than 90% as determined by SDS-PAGE. |
20300 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |