missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse BTNL2/Butyrophilin-like 2 Protein

Product Code. 18705007
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18705007

Brand: R&D Systems™ 8605BT050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Learn More About B7-related Butryophilins as Immune Modulators View Webinar

Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 1-6 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O70355
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 56244
Molecular Weight (g/mol) M.W. (Observed): 54-67 kDa, reducing conditions; M.W. (theoretical): 49 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse BTNL2/Butyrophilin-like 2 protein Asp27-Ser452, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias BTL-II, Butyrophilin like 2, Butyrophilin-like 2, butyrophilin-like 2 (MHC class II associated)
Common Name BTNL2/Butyrophilin-like 2
Gene Symbol Btnl2
Biological Activity Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is typically 1-6μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Expression System Mouse myeloma cell line, NS0-derived Asp27-Ser452, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein BTNL2/Butyrophilin-like 2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.