missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BMPR-IA/ALK-3 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£357.44
Specifications
Gene ID (Entrez) | 12166 |
---|---|
Molecular Weight (g/mol) | 41.5 kDa (monomer) |
Name | BMPR-IA/ALK-3 Fc Chimera |
Quantity | 100μg |
Source | Mouse myeloma cell line,NS0-derived mouse BMPR-IA/ALK-3 protein, (N-terminus) Mouse BMPR-IA (Gln24-Arg152) Accession &Num; Q60607 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18157360
|
R&D Systems™
437-MR-100 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£357.44
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18177670
|
R&D Systems™
437-MR-100/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£357.44
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse BMPR-IA/ALK-3 Fc Chimera Protein is derived from NS0. The Recombinant Mouse BMPR-IA/ALK-3 Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifications
12166 | |
BMPR-IA/ALK-3 Fc Chimera | |
Mouse myeloma cell line,NS0-derived mouse BMPR-IA/ALK-3 protein, (N-terminus) Mouse BMPR-IA (Gln24-Arg152) Accession &Num; Q60607 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
41.5 kDa (monomer) | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |