missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Betacellulin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1025-CE-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Mouse Betacellulin Protein is derived from E. coli. The Recombinant Mouse Betacellulin Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
9.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
12223 | |
Betacellulin/BTC | |
E. coli-derived mouse Betacellulin/BTC protein Asp32-Gln118 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |