missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Mouse beta-Synuclein Active, Monomer Protein
Shop All Bio Techne ProductsSpecifications
Specifications
| For Use With (Application) | SDS-PAGE, Western Blot |
| Formulation | PBS pH 7.4 |
| Gene ID (Entrez) | 6620 |
| Quantity | 100 μg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | beta-synuclein, synuclein, beta |
| Gene Symbol | SNCB |
| Research Category | Alzheimers Research, Neuroscience |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | beta-Synuclein |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?