missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse BD 3 Protein
A cDNA sequence encoding the BD 3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10532-1mg
Additional Details : Weight : 0.01000kg
Specifications
27358 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its antimicrobial activity against E. coli. The ED50 for this effect is typically 4-20microg/ml, corresponding to a specific activity of 50,000-250,000units/mg. | |
Mouse | |
Untagged | |
The Mouse BD-3 was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
BD 3 Protein | |
Research Use Only | |
Defb3 | |
Recombinant Protein | |
E. coli | |
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |