missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse BD 3 Protein

A cDNA sequence encoding the BD 3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10532-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954158

  • £155.00 / 20µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

27358
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Measured by its antimicrobial activity against E. coli. The ED50 for this effect is typically 4-20microg/ml, corresponding to a specific activity of 50,000-250,000units/mg.
Mouse
Untagged
The Mouse BD-3 was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
BD 3 Protein
Research Use Only
Defb3
Recombinant Protein
E. coli
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers