missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Bcl-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Bcl-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5710-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033871.2 | |
Bcl-2 Protein | |
Research Use Only | |
Bcl2 | |
Recombinant Protein | |
E. coli | |
GRTGYDNREI VMKYIHYKLS QRGYEWDAGD ADAAPLGAAP TPGIFSFQPE SNPMPAVHRD MAARTSPLRP LVATAGPALS PVPPVVHLTL RRAGDDFSRR YRRDFAEMSS QLHLTPFTAR GRFATVVEEL FRDGVNWGRI VAFFEFGGVM CVESVNREMS PLVDNIALWM TEYLNRHLHT WIQDNGGWDA FVELYGPSMR P | |
Greater than 90% as determined by SDS-PAGE. |
12043 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |