missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BAFF/BLyS/TNFSF13B Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Mouse BAFF/BLyS/TNFSF13B Protein is derived from NS0. The Recombinant Mouse BAFF/BLyS/TNFSF13B Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q9WU72 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 10673 |
| Molecular Weight (g/mol) | Observed M.W.: 19-23kDa, reducing conditions, Theoretical M.W.: 21kDa |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse BAFF/BLyS/TNFSF13B protein Ala127-Leu309 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4 |
| Common Name | BAFF/BLyS/TNFSF13B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction