missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse B7-H4 Fc Chimera Protein

Product Code. 18781098
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18781098 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18781098 Supplier R&D Systems™ Supplier No. 4206B701M
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit anti-CD3-induced proliferation of stimulated human T cells. Human T lymphocytes cultured for 72 hours with PHA were incubated for an additional 3 days in 96 well plates coated with 500 ng/mL anti-CD3 and Recombinant Mouse B7-H4 Fc Chimera. The ED50 for this effect is 0.5-3 μg/mL. Optimal dilutions should be determined by each laboratory for each application.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAP37284
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 79679
Molecular Weight (g/mol) M.W. (Observed): 75-85 kDa, reducing conditions; M.W. (theoretical): 52.4 kDa (monomer)
Quantity 1 mg
Source Mouse myeloma cell line, NS0-derived mouse B7-H4 protein Mouse B7-H4 (Phe29-Pro258) (N-terminus) Accession # AAP37284 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1
Gene Symbol Vtcn1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.