missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse B7-1/CD80 Protein

Product Code. 18490542 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
1 mg
2 mg
Unit Size:
0.05mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18490542 1 mg 1mg
18431342 2 mg 2mg
18719273 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18490542 Supplier Novus Biologicals™ Supplier No. NBP2265781mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page, Cell depletion

A recombinant protein corresponding to CD80. Amino Acid Sequence: MGHTRRQGTSPSKCPYLNFFQLLVLAGLSHFCSGVIHVTKEVKEVATL SCGHNVSVEELAQTRIYWQKEKKMVLTMMSGDMNIWPEYKNRTIFDI TNNLSIVILALRPSDEGTYECVVLKYEKDAFKREHLAEVTLSVKADFPT PSISDFEIPTSNIRRIICSTSGGFPEPHLSWLENGEELNAINTTVSQDPETEL YAVSSKLDFNMTTNHSFMCLIKYGHLRVNQTFNWNTTKQEHFPDDKTHT CPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYV DGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKV SNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDI AVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVM HEALHNHYTQKSLSLSPGK The Recombinant Mouse B7-1/CD80 Protein has been validated for the following applications: SDS-Page, Cell depletion.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 941
Name B7-1/CD80 Protein
Purification Method Protein A purified
Quantity 1 mg
Source Baculovirus
Storage Requirements -80°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.