missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein corresponding to CD80. Amino Acid Sequence: MGHTRRQGTSPSKCPYLNFFQLLVLAGLSHFCSGVIHVTKEVKEVATL SCGHNVSVEELAQTRIYWQKEKKMVLTMMSGDMNIWPEYKNRTIFDI TNNLSIVILALRPSDEGTYECVVLKYEKDAFKREHLAEVTLSVKADFPT PSISDFEIPTSNIRRIICSTSGGFPEPHLSWLENGEELNAINTTVSQDPETEL YAVSSKLDFNMTTNHSFMCLIKYGHLRVNQTFNWNTTKQEHFPDDKTHT CPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYV DGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKV SNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDI AVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVM HEALHNHYTQKSLSLSPGK The Recombinant Mouse B7-1/CD80 Protein has been validated for the following applications: SDS-Page, Cell depletion.
Specifications
Specifications
| Gene ID (Entrez) | 941 |
| Name | B7-1/CD80 Protein |
| Purification Method | Protein A purified |
| Quantity | 2 mg |
| Source | Baculovirus |
| Storage Requirements | -80°C. Avoid freeze-thaw cycles. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?