missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ATP synthase subunit beta, mitochondrial Protein

A DNA sequence encoding the Mus musculus (Mouse) ATP synthase subunit beta, mitochondrial, was expressed in the hosts and tags indicated.

£223.39 - £2475.95

Specifications

Name ATP synthase subunit beta, mitochondrial Protein
Product Type Recombinant Protein
Protein Tag His-SUMO
Host Species E. coli
Cross Reactivity Mouse
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15947059
View Documents Promotion Details
enQuireBio™
QP5694-EC-100UG
100 μg
£560.06
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-02-2022
Please to purchase this item. Need a web account? Register with us today!
15967059
View Documents Promotion Details
enQuireBio™
QP5694-EC-1MG
1 mg
£2475.95
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 18-02-2022
Please to purchase this item. Need a web account? Register with us today!
15957059
View Documents Promotion Details
enQuireBio™
QP5694-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15977059
View Documents Promotion Details
enQuireBio™
QP5694-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15987059
View Documents Promotion Details
enQuireBio™
QP5694-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15997059
View Documents Promotion Details
enQuireBio™
QP5694-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

ATP synthase subunit beta, mitochondrial Protein
His-SUMO
Mouse
Greater than 90% as determined by SDS-PAGE.
NP_058054.2
Recombinant Protein
E. coli
AAQASAAPKA GTATGRIVAV IGAVVDVQFD EGLPPILNAL EVQGRDSRLV LEVAQHLGES TVRTIAMDGT EGLVRGQKVL DSGAPIKIPV GPETLGRIMN VIGEPIDERG PIKTKQFAPI HAEAPEFIEM SVEQEILVTG IKVVDLLAPY AKGGKIGLFG GAGVGKTVLI MELINNVAKA HGGYSVFAGV GERTREGNDL YHEMIESGVI NLKDATSKVA LVYGQMNEPP GARARVALTG LTVAEYFRDQ EGQDVLLFID NIFRFTQAGS EVSALLGRIP SAVGYQPTLA TDMGTMQERI TTTKKGSITS VQAIYVPADD LTDPAPATTF AHLDATTVLS RAIAELGIYP AVDPLDSTSR IMDPNIVGNE HYDVARGVQK ILQDYKSLQD IIAILGMDEL SEEDKLTVSR ARKIQRFLSQ PFQVAEVFTG HMGKLVPLKE TIKGFQQILA GEYDHLPEQA FYMVGPIEEA VAKADKLAEE HGS
11947
Atp5b
SDS
Product Certifications