missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse APRIL/TNFSF13 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
TNFSF13 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using activated T cells.
Specifications
Specifications
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, with 0.02 % Tween-20. |
| Gene ID (Entrez) | 8741 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 16.4 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of APRIL/TNFSF13 as determined by LAL method. |
| Gene Alias | A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, sAPRIL, sAPRIL/TNFSF13, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 |
| Gene Symbol | TNFSF13 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction