missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ANTXR1 Protein
A DNA sequence encoding the Mus musculus (Mouse) ANTXR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5649-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_473382.1 | |
ANTXR1 Protein | |
Research Use Only | |
Antxr1 | |
Recombinant Protein | |
Yeast | |
EDGGPACYGG FDLYFILDKS GSVLHHWNEI YYFVEQLAHR FISPQLRMSF IVFSTRGTTL MKLTEDREQI RQGLEELQKV LPGGDTYMHE GFERASEQIY YENSQGYRTA SVIIALTDGE LHEDLFFYSE REANRSRDLG AIVYCVGVKD FNETQLARIA DSKDHVFPVN DGFQALQGII HSILKKSCIE ILAAEPSTIC AGESFQVVVR GNGFRHARNV DRVLCSFKIN DSVTLNEKPF AVEDTYLLCP APILKEVGMK AALQVSMNDG LSFISSSVII TTTHCSDGS | |
Greater than 90% as determined by SDS-PAGE. |
69538 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |