missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ANGPTL7 Protein
A cDNA sequence encoding the ANGPTL7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11022-25ug
Additional Details : Weight : 0.01000kg
Specifications
ANGPTL7 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
QKPHKRKTQL KAAGCCEEMR ELKAQVANLS SLLGELSRKQ ESDWVSVVMQ VMELESSSKH MESRLSTAES KYSEMNNQID IMQLQAAQTV TQTSADAIYD CSSLYQKNYR ISGVYKLPPD EFLGSPELEV FCDMETSGGG WTIIQRRKSG LVSFYQDWRQ YKQGFGSIRG DFWLGNEHIH RLTRQPSRLR VELEDWEGNA RYAEYSYFAL GNELNSYRLF LGNYSGNVGK DALLYHNNTV FSTKDKDNDN CLDKCAQLRK GGYWYNCCTD SNLNGVYYRL GEHRKHMDGI SWYGWHGANY SLKRVEMKIR PEAFKPLEHH HHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
ANGPTL7 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |